Assay Details
Gene Symbol: Krit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170552.1 | NP_001164023.1 | 10 - 11 | 1613 | 91 | |
NM_030675.3 | NP_109600.2 | 10 - 11 | 1567 | 91 | ||
NR_033173.1 | 11 - 12 | 1814 | 91 | |||
GenBank mRNA | AF306509.1 | - | 10 - 11 | 1262 | 91 | |
AF310134.1 | - | 10 - 11 | 1543 | 91 | ||
AK039264.1 | - | 1444 | 91 | |||
AK085107.1 | - | 10 - 11 | 1595 | 91 | ||
AK144667.1 | - | 10 - 11 | 1549 | 91 | ||
AY328895.1 | - | 9 - 10 | 1249 | 91 | ||
AY464945.1 | - | 9 - 10 | 1249 | 91 | ||
BC016087.1 | - | 10 - 11 | 1517 | 91 | ||
BC044820.1 | - | 10 - 11 | 1456 | 91 | ||
BC054819.1 | - | 11 - 12 | 1806 | 91 | ||
BC085252.1 | - | 10 - 11 | 1373 | 91 | ||
BC141042.1 | - | 10 - 11 | 1411 | 9055654 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
