Assay Details
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010113.3 | NP_034243.2 | 14 - 15 | 2596 | 67 | |
GenBank mRNA | AK085707.1 | - | 14 - 15 | 2163 | 67 | |
AK088408.1 | - | 9 - 10 | 1183 | 67 | ||
AK136439.1 | - | 9 - 10 | 1189 | 67 | ||
AK142666.1 | - | 14 - 15 | 2292 | 67 | ||
BC017681.1 | - | 15 - 16 | 2533 | 4221592 | 67 | |
BC060741.1 | - | 14 - 15 | 2581 | 4164289 | 67 | |
BC092277.1 | - | 14 - 15 | 2581 | 4240429 | 67 | |
J00380.1 | - | 14 - 15 | 2591 | 67 | ||
V00741.1 | - | 14 - 15 | 2547 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
