Assay Details
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010113.3 | NP_034243.2 | 5 - 6 | 1313 | 69 | |
GenBank mRNA | AK085707.1 | - | 5 - 6 | 880 | 69 | |
AK142666.1 | - | 5 - 6 | 1009 | 69 | ||
BC017681.1 | - | 6 - 7 | 1250 | 4221592 | 69 | |
BC060741.1 | - | 5 - 6 | 1298 | 4164289 | 69 | |
BC092277.1 | - | 5 - 6 | 1298 | 4240429 | 69 | |
J00380.1 | - | 5 - 6 | 1308 | 69 | ||
V00741.1 | - | 5 - 6 | 1264 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
