Assay Details
Gene Symbol: Ebf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276387.1 | NP_001263316.1 | 1 - 2 | 667 | 66 | |
GenBank mRNA | AK139262.1 | - | 1 - 2 | 374 | 66 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK139262.1, NM_001276387.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
