Assay Details
Gene Symbol: Dpyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170778.2 | NP_740748.1 | 3 - 4 | 334 | 57 | |
GenBank mRNA | AK145094.1 | - | 3 - 4 | 382 | 57 | |
AK149426.1 | - | 3 - 4 | 336 | 57 | ||
BC039699.1 | - | 3 - 4 | 335 | 4235971 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
