Assay Details
Gene Symbol: Prc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285997.1 | NP_001272926.1 | 2 - 3 | 272 | 76 | |
NM_001285998.1 | NP_001272927.1 | 2 - 3 | 272 | 76 | ||
NM_145150.3 | NP_660132.2 | 2 - 3 | 272 | 76 | ||
GenBank mRNA | AK029140.1 | - | 2 - 3 | 259 | 76 | |
AK081506.1 | - | 2 - 3 | 245 | 76 | ||
AK088576.1 | - | 2 - 3 | 244 | 76 | ||
BC005475.1 | - | 2 - 3 | 206 | 3592138 | 76 | |
BC059808.1 | - | 2 - 3 | 217 | 76 | ||
CF736005.1 | - | 2 - 3 | 272 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
