Assay Details
Gene Symbol: 2900076A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045299.1 | 2 - 3 | 285 | 82 | ||
GenBank mRNA | AK013794.1 | - | 1 - 2 | 151 | 82 | |
BB620528.1 | - | 2 - 3 | 266 | 82 | ||
CF168906.1 | - | 2 - 3 | 283 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cDNA sequence BC048679 | BC048679 | -30.5kb | Anti-sense | Chr. 7 - 81494272 - 81498330 on NCBI Build GRCm38 | |
Downstream | microRNA 1839 | Mir1839 |
mir-1839, mmu-mir-1839 |
482.0 | Sense | Chr. 7 - 81529916 - 81529988 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
