Assay Details
Gene Symbol: Furin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081454.1 | NP_001074923.1 | 4 - 5 | 649 | 78 | |
NM_011046.2 | NP_035176.1 | 4 - 5 | 769 | 78 | ||
GenBank mRNA | AK199186.1 | - | 1 - 2 | 55 | 78 | |
BC048234.1 | - | 3 - 4 | 476 | 6492009 | 78 | |
L26489.1 | - | 4 - 5 | 769 | 78 | ||
X54056.1 | - | 3 - 4 | 472 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
