Assay Details
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281929.1 | NP_001268858.1 | 10 - 11 | 1745 | 93 | |
NM_001290451.1 | NP_001277380.1 | 10 - 11 | 1517 | 93 | ||
NM_016886.4 | NP_058582.3 | 10 - 11 | 1745 | 93 | ||
NR_104053.1 | 10 - 11 | 1745 | 93 | |||
GenBank mRNA | AB022342.1 | - | 10 - 11 | 1535 | 93 | |
AB079072.1 | - | 10 - 11 | 1481 | 93 | ||
AB102775.1 | - | 10 - 11 | 1481 | 93 | ||
AB102776.1 | - | 10 - 11 | 1481 | 93 | ||
AF483494.1 | - | 10 - 11 | 1481 | 93 | ||
AF483495.1 | - | 10 - 11 | 1481 | 93 | ||
AK135043.1 | - | 10 - 11 | 1499 | 93 | ||
AK138615.1 | - | 10 - 11 | 1650 | 93 | ||
AK157815.1 | - | 11 - 12 | 1922 | 93 | ||
AK161163.1 | - | 10 - 11 | 1565 | 93 | ||
AK220530.1 | - | 10 - 11 | 1532 | 93 | ||
BC076584.1 | - | 10 - 11 | 1609 | 93 | ||
BC086678.1 | - | 10 - 11 | 1690 | 30668476 | 93 | |
BC129855.1 | - | 10 - 11 | 1535 | 40109751 | 93 | |
BC129856.1 | - | 10 - 11 | 1535 | 40109752 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
