Assay Details
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281929.1 | NP_001268858.1 | 4 - 5 | 936 | 131 | |
NM_001290451.1 | NP_001277380.1 | 4 - 5 | 708 | 131 | ||
NM_016886.4 | NP_058582.3 | 4 - 5 | 936 | 131 | ||
NR_104053.1 | 4 - 5 | 936 | 131 | |||
GenBank mRNA | AB022342.1 | - | 4 - 5 | 726 | 131 | |
AB079072.1 | - | 4 - 5 | 672 | 131 | ||
AB102775.1 | - | 4 - 5 | 672 | 131 | ||
AB102776.1 | - | 4 - 5 | 672 | 131 | ||
AF483494.1 | - | 4 - 5 | 672 | 131 | ||
AF483495.1 | - | 4 - 5 | 672 | 131 | ||
AK135043.1 | - | 4 - 5 | 689 | 131 | ||
AK138615.1 | - | 4 - 5 | 841 | 131 | ||
AK157815.1 | - | 5 - 6 | 1113 | 131 | ||
AK161163.1 | - | 4 - 5 | 756 | 131 | ||
AK220530.1 | - | 4 - 5 | 723 | 131 | ||
BC076584.1 | - | 4 - 5 | 801 | 131 | ||
BC086678.1 | - | 4 - 5 | 881 | 30668476 | 131 | |
BC129855.1 | - | 4 - 5 | 726 | 40109751 | 131 | |
BC129856.1 | - | 4 - 5 | 726 | 40109752 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
