Assay Details
Gene Symbol: Gria3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281929.1 | NP_001268858.1 | 8 - 9 | 1436 | 140 | |
NM_001290451.1 | NP_001277380.1 | 8 - 9 | 1208 | 140 | ||
NM_016886.4 | NP_058582.3 | 8 - 9 | 1436 | 140 | ||
NR_104053.1 | 8 - 9 | 1436 | 140 | |||
GenBank mRNA | AB022342.1 | - | 8 - 9 | 1226 | 140 | |
AB079072.1 | - | 8 - 9 | 1172 | 140 | ||
AB102775.1 | - | 8 - 9 | 1172 | 140 | ||
AB102776.1 | - | 8 - 9 | 1172 | 140 | ||
AF483494.1 | - | 8 - 9 | 1172 | 140 | ||
AF483495.1 | - | 8 - 9 | 1172 | 140 | ||
AK138615.1 | - | 8 - 9 | 1341 | 140 | ||
AK157815.1 | - | 9 - 10 | 1613 | 140 | ||
AK161163.1 | - | 8 - 9 | 1256 | 140 | ||
AK220530.1 | - | 8 - 9 | 1223 | 140 | ||
BC076584.1 | - | 8 - 9 | 1300 | 139 | ||
BC086678.1 | - | 8 - 9 | 1381 | 30668476 | 140 | |
BC129855.1 | - | 8 - 9 | 1226 | 40109751 | 140 | |
BC129856.1 | - | 8 - 9 | 1226 | 40109752 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
