Assay Details
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033272.1 | NP_001028444.1 | 3 - 4 | 2199 | 83 | |
GenBank mRNA | BC035942.1 | - | 1 - 2 | 218 | 83 | |
BC138455.1 | - | 3 - 4 | 2156 | 8861475 | 83 | |
BC145285.1 | - | 3 - 4 | 2156 | 9053839 | 83 | |
CN455410.1 | - | 2 - 3 | 594 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
