Assay Details
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033272.1 | NP_001028444.1 | 9 - 10 | 3255 | 112 | |
GenBank mRNA | AK151394.1 | - | 6 - 7 | 1136 | 112 | |
AK158193.1 | - | 1 - 2 | 789 | 112 | ||
AK220313.1 | - | 6 - 7 | 977 | 112 | ||
BC035942.1 | - | 7 - 8 | 1273 | 112 | ||
BC138455.1 | - | 9 - 10 | 3212 | 8861475 | 112 | |
BC145285.1 | - | 9 - 10 | 3212 | 9053839 | 112 | |
CN538317.1 | - | 3 - 4 | 437 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
