Assay Details
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033272.1 | NP_001028444.1 | 10 - 11 | 3395 | 65 | |
GenBank mRNA | AK151394.1 | - | 7 - 8 | 1276 | 65 | |
AK158193.1 | - | 2 - 3 | 929 | 65 | ||
AK220313.1 | - | 7 - 8 | 1117 | 65 | ||
BC035942.1 | - | 8 - 9 | 1413 | 65 | ||
BC138455.1 | - | 10 - 11 | 3352 | 8861475 | 65 | |
BC145285.1 | - | 10 - 11 | 3352 | 9053839 | 65 | |
CN538317.1 | - | 4 - 5 | 577 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
