Assay Details
Gene Symbol: 1700015F17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001200025.1 | NP_001186954.1 | 6 - 7 | 622 | 84 | |
GenBank mRNA | AK005993.1 | - | 6 - 7 | 572 | 84 | |
BC052364.1 | - | 7 - 8 | 967 | 84 | ||
BC099562.1 | - | 6 - 7 | 581 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin-dependent kinase 14 | Cdk14 |
Pftk1, mKIAA0834 |
-57.6kb | Sense | Chr. 5 - 4803384 - 5380251 on NCBI Build GRCm38 |
Downstream | claudin 12 | Cldn12 | 54.4kb | Sense | Chr. 5 - 5505015 - 5514976 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132695 | NM_001200025.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132695 | AK005993.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132695 | AV044423.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P132695 | BC099562.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132695 | NM_001200025.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132695 | AK005993.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132695 | AV044423.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P132695 | BC099562.1 | mouse_transcriptome |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455978 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04485682 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04897118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04914880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04797024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455905 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04877510 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04767807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455910 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04524314 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455789 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455969 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04360202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04197713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04476462 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04767808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23456001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455765 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455776 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455927 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455820 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23456000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04197701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455996 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04550024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04693922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04846878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04851582 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04610757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455786 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455872 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04721215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04987029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04914882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04476459 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04626524 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04360204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04114363 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04485678 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455987 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04877505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455816 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455829 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455991 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455768 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455897 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455828 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23455961 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1456366_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456366_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456366_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381716_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381716_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456366_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1456366_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217218 | - | - |
