Assay Details
Gene Symbol: Acin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085473.2 | NP_001078942.2 | 6 - 7 | 2112 | 67 | |
NM_023190.3 | NP_075679.2 | 6 - 7 | 2112 | 67 | ||
GenBank mRNA | AF168782.1 | - | 6 - 7 | 2113 | 67 | |
AK030449.1 | - | 6 - 7 | 1805 | 67 | ||
AK050467.1 | - | 5 - 6 | 1675 | 67 | ||
AK122342.1 | - | 6 - 7 | 1783 | 67 | ||
BC075691.1 | - | 6 - 7 | 1797 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
