Assay Details
Gene Symbol: Abca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007378.1 | NP_031404.1 | 16 - 17 | 2672 | 74 | |
GenBank mRNA | AF000149.1 | - | 16 - 17 | 2672 | 74 | |
BC057853.1 | - | 16 - 17 | 2672 | 6492139 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
