Assay Details
Gene Symbol: Tank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164071.1 | NP_001157543.1 | 4 - 5 | 572 | 91 | |
NM_001164072.1 | NP_001157544.1 | 3 - 4 | 364 | 91 | ||
NM_011529.2 | NP_035659.1 | 3 - 4 | 363 | 91 | ||
GenBank mRNA | AK031559.1 | - | 3 - 4 | 326 | 91 | |
AK088817.1 | - | 3 - 4 | 310 | 91 | ||
AK147727.1 | - | 4 - 5 | 518 | 91 | ||
AK165420.1 | - | 3 - 4 | 320 | 91 | ||
AK165994.1 | - | 3 - 4 | 295 | 91 | ||
BB660164.1 | - | 3 - 4 | 365 | 91 | ||
BC026148.1 | - | 3 - 4 | 345 | 3492937 | 91 | |
BC033282.1 | - | 3 - 4 | 299 | 91 | ||
BC052685.1 | - | 3 - 4 | 264 | 91 | ||
U51907.1 | - | 3 - 4 | 302 | 91 | ||
U59864.1 | - | 3 - 4 | 376 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
