Assay Details
Gene Symbol: Ddx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033294.3 | NP_001028466.2 | 7 - 8 | 1017 | 78 | |
GenBank mRNA | AK030888.1 | - | 7 - 8 | 964 | 78 | |
AK144739.1 | - | 7 - 8 | 730 | 78 | ||
BC066017.1 | - | 7 - 8 | 654 | 6401415 | 78 | |
BC132351.1 | - | 7 - 8 | 707 | 40130628 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
