Assay Details
Gene Symbol: Begain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163175.1 | NP_001156647.1 | 4 - 5 | 350 | 63 | |
GenBank mRNA | AK158915.1 | - | 4 - 5 | 351 | 63 | |
BC080282.1 | - | 2 - 3 | 143 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
