Assay Details
Gene Symbol: Kif16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081133.2 | NP_001074602.1 | 15 - 16 | 1705 | 98 | |
GenBank mRNA | AB610968.1 | - | 15 - 16 | 1760 | 98 | |
AK033018.1 | - | 9 - 10 | 1032 | 98 | ||
AK129401.1 | - | 2 - 3 | 156 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
