Assay Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244891.1 | NP_001231820.1 | 9 - 10 | 1070 | 84 | |
NM_001244903.1 | NP_001231832.1 | 8 - 9 | 1017 | 84 | ||
NM_017368.3 | NP_059064.2 | 9 - 10 | 1169 | 84 | ||
NM_198683.2 | NP_941955.1 | 8 - 9 | 1017 | 84 | ||
GenBank mRNA | AB050499.1 | - | 5 - 6 | 478 | 84 | |
AF267535.1 | - | 7 - 8 | 773 | 84 | ||
AF314172.1 | - | 7 - 8 | 611 | 84 | ||
AJ007987.1 | - | 7 - 8 | 762 | 84 | ||
AK014492.1 | - | 8 - 9 | 930 | 84 | ||
AK143698.1 | - | 9 - 10 | 971 | 84 | ||
AK156725.1 | - | 8 - 9 | 930 | 84 | ||
AK211397.1 | - | 3 - 4 | 281 | 84 | ||
AK218377.1 | - | 4 - 5 | 342 | 84 | ||
BC021393.1 | - | 8 - 9 | 882 | 84 | ||
X61451.1 | - | 4 - 5 | 465 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
