Assay Details
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244891.1 | NP_001231820.1 | 12 - 13 | 1502 | 71 | |
NM_001244903.1 | NP_001231832.1 | 11 - 12 | 1449 | 71 | ||
NM_017368.3 | NP_059064.2 | 12 - 13 | 1601 | 71 | ||
NM_198683.2 | NP_941955.1 | 11 - 12 | 1449 | 71 | ||
GenBank mRNA | AB050499.1 | - | 8 - 9 | 913 | 71 | |
AF267535.1 | - | 10 - 11 | 1205 | 71 | ||
AF314172.1 | - | 10 - 11 | 1046 | 71 | ||
AJ007987.1 | - | 10 - 11 | 1194 | 71 | ||
AK014492.1 | - | 11 - 12 | 1365 | 71 | ||
AK143698.1 | - | 12 - 13 | 1403 | 71 | ||
AK183548.1 | - | 2 - 3 | 208 | 71 | ||
AK188588.1 | - | 2 - 3 | 208 | 71 | ||
AK200025.1 | - | 1 - 2 | 157 | 71 | ||
BC021393.1 | - | 11 - 12 | 1314 | 71 | ||
CF171545.1 | - | 3 - 4 | 71 | |||
CF745326.1 | - | 1 - 2 | 139 | 71 | ||
X61451.1 | - | 7 - 8 | 899 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
