Assay Details
Gene Symbol: Tlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172664.3 | NP_766252.2 | 6 - 7 | 702 | 72 | |
GenBank mRNA | AK029773.1 | - | 5 - 6 | 428 | 72 | |
BC033931.1 | - | 4 - 5 | 264 | 72 | ||
BC051641.1 | - | 6 - 7 | 702 | 6507968 | 72 | |
BC057369.1 | - | 7 - 8 | 688 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
