Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200031358.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031358.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031358.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031362.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031362.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031362.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031348.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031348.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031348.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031350.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031350.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031350.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031351.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031351.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031351.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031353.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031353.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031353.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031360.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031360.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031360.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031349.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031349.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031349.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031356.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031356.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031356.mm.1 |
NM_018733.2 |
Refseq |