Assay Details
Gene Symbol: Stxbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113569.1 | NP_001107041.1 | 10 - 11 | 1044 | 87 | |
NM_009295.2 | NP_033321.2 | 10 - 11 | 1039 | 87 | ||
GenBank mRNA | AF326545.1 | - | 10 - 11 | 898 | 87 | |
AF326563.1 | - | 10 - 11 | 898 | 87 | ||
AK004780.1 | - | 5 - 6 | 763 | 87 | ||
BC031728.1 | - | 10 - 11 | 971 | 4480978 | 87 | |
D45903.1 | - | 10 - 11 | 933 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
