Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290413.1 | NP_001277342.1 | 3 - 4 | 396 | 79 | |
NM_009422.3 | NP_033448.2 | 3 - 4 | 375 | 79 | ||
GenBank mRNA | AF027570.1 | - | 2 - 3 | 287 | 79 | |
AK038136.1 | - | 3 - 4 | 324 | 79 | ||
AK052934.1 | - | 3 - 4 | 324 | 79 | ||
AK151653.1 | - | 3 - 4 | 313 | 79 | ||
AK152174.1 | - | 3 - 4 | 343 | 79 | ||
AK159581.1 | - | 3 - 4 | 345 | 79 | ||
AK159585.1 | - | 322 | 79 | |||
AK184736.1 | - | 2 - 3 | 112 | 79 | ||
BC003801.1 | - | 2 - 3 | 301 | 3492839 | 79 | |
BC060625.1 | - | 2 - 3 | 352 | 79 | ||
L35303.1 | - | 3 - 4 | 312 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
