Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290413.1 | NP_001277342.1 | 5 - 6 | 651 | 80 | |
NM_009422.3 | NP_033448.2 | 5 - 6 | 630 | 80 | ||
GenBank mRNA | AF027570.1 | - | 4 - 5 | 542 | 80 | |
AK038136.1 | - | 5 - 6 | 579 | 80 | ||
AK151653.1 | - | 5 - 6 | 568 | 80 | ||
AK152174.1 | - | 5 - 6 | 598 | 80 | ||
AK159581.1 | - | 5 - 6 | 600 | 80 | ||
AK159585.1 | - | 577 | 80 | |||
BC003801.1 | - | 4 - 5 | 556 | 3492839 | 80 | |
L35303.1 | - | 5 - 6 | 567 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
