Assay Details
Gene Symbol: Ambra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080754.1 | NP_001074223.1 | 13 - 14 | 2726 | 102 | |
NM_172669.3 | NP_766257.3 | 12 - 13 | 2999 | 102 | ||
GenBank mRNA | AK129433.1 | - | 8 - 9 | 2263 | 102 | |
AK148887.1 | - | 12 - 13 | 2631 | 102 | ||
AK155513.1 | - | 12 - 13 | 2962 | 102 | ||
AK169280.1 | - | 13 - 14 | 2679 | 102 | ||
BC032293.1 | - | 13 - 14 | 2649 | 102 | ||
BC054082.1 | - | 12 - 13 | 2932 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
