Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009396.2 | NP_033422.2 | 2 - 3 | 380 | 60 | |
GenBank mRNA | AK087680.1 | - | 2 - 3 | 379 | 60 | |
AK154155.1 | - | 2 - 3 | 383 | 60 | ||
AK154405.1 | - | 2 - 3 | 380 | 60 | ||
AK170719.1 | - | 2 - 3 | 380 | 60 | ||
AK172653.1 | - | 2 - 3 | 380 | 60 | ||
AK191831.1 | - | 1 - 2 | 182 | 60 | ||
BC138106.1 | - | 1 - 2 | 289 | 8861125 | 60 | |
BC138107.1 | - | 1 - 2 | 289 | 8861126 | 60 | |
L24118.1 | - | 1 - 2 | 324 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
