Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009396.2 | NP_033422.2 | 4 - 5 | 1125 | 54 | |
GenBank mRNA | AK087680.1 | - | 4 - 5 | 1126 | 54 | |
AK154155.1 | - | 4 - 5 | 1128 | 54 | ||
AK154405.1 | - | 4 - 5 | 1125 | 54 | ||
AK170719.1 | - | 4 - 5 | 1125 | 54 | ||
AK172653.1 | - | 4 - 5 | 1125 | 54 | ||
BC138106.1 | - | 3 - 4 | 1034 | 8861125 | 54 | |
BC138107.1 | - | 3 - 4 | 1034 | 8861126 | 54 | |
L24118.1 | - | 3 - 4 | 1069 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
