Assay Details
Gene Symbol: Tnfaip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009396.2 | NP_033422.2 | 6 - 7 | 1343 | 102 | |
GenBank mRNA | AK154155.1 | - | 6 - 7 | 1397 | 102 | |
AK154405.1 | - | 6 - 7 | 1343 | 102 | ||
AK170719.1 | - | 6 - 7 | 1343 | 102 | ||
AK172653.1 | - | 6 - 7 | 1343 | 102 | ||
BC138106.1 | - | 5 - 6 | 1252 | 8861125 | 102 | |
BC138107.1 | - | 5 - 6 | 1252 | 8861126 | 102 | |
L24118.1 | - | 5 - 6 | 1287 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
