Assay Details
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010726.2 | NP_034856.1 | 8 - 9 | 1038 | 91 | |
GenBank mRNA | AF023463.1 | - | 8 - 9 | 1003 | 91 | |
AK146753.1 | - | 8 - 9 | 1023 | 91 | ||
AK151372.1 | - | 8 - 9 | 986 | 91 | ||
AK164495.1 | - | 8 - 9 | 989 | 91 | ||
AK165412.1 | - | 8 - 9 | 982 | 91 | ||
AK168648.1 | - | 8 - 9 | 1008 | 91 | ||
AK169213.1 | - | 8 - 9 | 980 | 91 | ||
BC002018.1 | - | 8 - 9 | 1015 | 3489939 | 91 | |
D88670.1 | - | 8 - 9 | 998 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276063 | AF023463.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P276063 | AF023463.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164908 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90563 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33125 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921375 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182267 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 447703 | AK085506.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348464 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 302573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76580 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610474 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 174847 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 147428 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P105360 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111235 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276063 | AF023463.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P276063 | AF023463.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276063 | AF023463.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P276063 | AF023463.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970972 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970991 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970968 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970989 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970969 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970980 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20970982 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04414589 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P105360 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04414602 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111235 | - | - |
Whole Mouse Genome G4122A | A_51_P276063 | AF023463.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P276063 | AF023463.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133293 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111235 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P105360 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02244597 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02244602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970968 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20970954 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20971076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02277375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01388420 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01937030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01372433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01643673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01556054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01250536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01530613 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971014 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01372431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01192633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02110196 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01192631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970968 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01336500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970994 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971012 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02263633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971108 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02244602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01512907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971115 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970986 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970991 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20970966 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971000 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20971001 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10469052 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469048 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469047 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469049 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469053 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469050 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469051 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469054 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10469055 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1460194_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10469046 | - | - |
Mouse Genome 430 2.0 Array | 1460194_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1460194_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16922_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16922_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200000306.mm.1 | TC0200000037.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200000306.mm.1 | NM_010726.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200000306.mm.1 | NM_010726.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200000305.mm.1 | TC0200000037.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200000305.mm.1 | NM_010726.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200000305.mm.1 | NM_010726.2 | Refseq |
Mouse Expression Set 430 | 1460194_at | - | - |
Mouse Expression Set 430 | 1460194_PM_at | - | - |
Murine Genome U74v2 Set | 96608_PM_at | - | - |
Murine Genome U74v2 Set | 96608_at | - | - |
Mouse Genome 430A 2.0 Array | 1460194_at | - | - |
Mouse Genome 430A 2.0 Array | 1460194_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460194_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1460194_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6380128 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6380128 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006380128 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2756238 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2756238 | - | - |
mouse-6 v1.1 (Illumina) | scl21324.9.1_204-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6380128 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl21324.9.1_204-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6380128 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2756238 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2756238 | - | - |
