Assay Details
Gene Symbol: Gas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008087.2 | NP_032113.1 | 5 - 6 | 719 | 144 | |
GenBank mRNA | AK014509.1 | - | 7 - 8 | 1224 | 144 | |
AK142903.1 | - | 7 - 8 | 898 | 144 | ||
BC013456.1 | - | 4 - 5 | 487 | 4237356 | 144 | |
BC053446.1 | - | 5 - 6 | 768 | 6336752 | 144 | |
M21828.1 | - | 5 - 6 | 719 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
