Assay Details
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172666.3 | NP_766254.2 | 11 - 12 | 1301 | 86 | |
GenBank mRNA | AK031049.1 | - | 11 - 12 | 1303 | 86 | |
AK031154.1 | - | 11 - 12 | 1142 | 86 | ||
AK042745.1 | - | 11 - 12 | 1141 | 86 | ||
AK053686.1 | - | 11 - 12 | 1290 | 86 | ||
AK079447.1 | - | 10 - 11 | 981 | 86 | ||
BC063086.1 | - | 11 - 12 | 1286 | 6332574 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
