Assay Details
Gene Symbol: Agps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172666.3 | NP_766254.2 | 9 - 10 | 1068 | 107 | |
GenBank mRNA | AK031049.1 | - | 9 - 10 | 1070 | 107 | |
AK031154.1 | - | 9 - 10 | 909 | 107 | ||
AK042745.1 | - | 9 - 10 | 908 | 107 | ||
AK053686.1 | - | 9 - 10 | 1057 | 107 | ||
AK079447.1 | - | 8 - 9 | 748 | 107 | ||
BC063086.1 | - | 9 - 10 | 1053 | 6332574 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
