Assay Details
Gene Symbol: Commd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147778.3 | NP_680087.1 | 2 - 3 | 248 | 72 | |
GenBank mRNA | AK007318.1 | - | 2 - 3 | 253 | 72 | |
AK041634.1 | - | 2 - 3 | 249 | 72 | ||
AK145955.1 | - | 2 - 3 | 276 | 72 | ||
AK145999.1 | - | 2 - 3 | 278 | 72 | ||
AK163947.1 | - | 1 - 2 | 1546 | 72 | ||
BC038077.1 | - | 2 - 3 | 290 | 4013160 | 72 | |
BC048815.1 | - | 2 - 3 | 259 | 3482368 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
