Assay Details
Gene Symbol: Dbhos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040524.1 | 6 - 7 | 1414 | 75 | ||
GenBank mRNA | AK032445.1 | - | 6 - 7 | 1416 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
family with sequence similarity 163, member B |
Fam163b | C630035N08Rik | -2.4kb | Sense | Chr. 2 - 27110379 - 27142477 on NCBI Build GRCm38 |
Downstream | dopamine beta hydroxylase | Dbh | 12.6kb | Anti-sense | Chr. 2 - 27165507 - 27183204 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
