Assay Details
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080853.3 | NP_543129.3 | 7 - 8 | 1692 | 83 | |
GenBank mRNA | AF324864.1 | - | 7 - 8 | 1525 | 83 | |
AK043753.1 | - | 6 - 7 | 1145 | 83 | ||
AK045409.1 | - | 7 - 8 | 1455 | 83 | ||
BC038375.1 | - | 7 - 8 | 1071 | 5357143 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
