Assay Details
Gene Symbol: Tmem87b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028248.2 | NP_082524.2 | 14 - 15 | 1413 | 121 | |
GenBank mRNA | AK011959.1 | - | 14 - 15 | 1358 | 121 | |
AK050203.1 | - | 14 - 15 | 1405 | 121 | ||
AK171811.1 | - | 15 - 16 | 1329 | 121 | ||
AK172611.1 | - | 15 - 16 | 1329 | 121 | ||
BC031488.1 | - | 14 - 15 | 1369 | 121 | ||
CN458042.1 | - | 6 - 7 | 548 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
