Assay Details
Gene Symbol: Slc43a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021398.3 | NP_067373.2 | 6 - 7 | 901 | 69 | |
GenBank mRNA | AF188622.1 | - | 6 - 7 | 895 | 69 | |
AK086962.1 | - | 5 - 6 | 1330 | 69 | ||
BC089603.1 | - | 5 - 6 | 601 | 6758493 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
