Assay Details
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081349.2 | NP_001074818.2 | 1 - 2 | 270 | 55 | |
GenBank mRNA | AB103034.1 | - | 1 - 2 | 239 | 55 | |
AK011417.1 | - | 1 - 2 | 269 | 55 | ||
AK046758.1 | - | 1 - 2 | 262 | 55 | ||
AK165374.1 | - | 1 - 2 | 262 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB103034.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
