Assay Details
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089281.1 | - | 1 - 2 | 55 | 61 | |
AK142931.1 | - | 1 - 2 | 70 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK089281.1, AK142931.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
