Assay Details
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009421.3 | NP_033447.2 | 7 - 8 | 759 | 79 | |
GenBank mRNA | AK029135.1 | - | 7 - 8 | 761 | 79 | |
AK089281.1 | - | 6 - 7 | 503 | 79 | ||
AK142931.1 | - | 6 - 7 | 518 | 79 | ||
AK145075.1 | - | 334 | 79 | |||
AK169515.1 | - | 7 - 8 | 552 | 79 | ||
AK172598.1 | - | 3 - 4 | 363 | 79 | ||
BC096595.1 | - | 3 - 4 | 325 | 30748947 | 79 | |
L35302.1 | - | 6 - 7 | 505 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
