Assay Details
Gene Symbol: Ppp1r12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029834.3 | NP_084110.2 | 2 - 3 | 543 | 73 | |
GenBank mRNA | AK010836.1 | - | 2 - 3 | 529 | 73 | |
AK046570.1 | - | 2 - 3 | 513 | 73 | ||
AK144521.1 | - | 2 - 3 | 487 | 73 | ||
AK144691.1 | - | 2 - 3 | 544 | 73 | ||
AK187068.1 | - | 2 - 3 | 154 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
