Assay Details
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177608.3 | NP_808276.2 | 12 - 13 | 1969 | 150 | |
GenBank mRNA | AK145135.1 | - | 12 - 13 | 1855 | 150 | |
AK172920.1 | - | 12 - 13 | 1836 | 150 | ||
BC071239.1 | - | 5 - 6 | 597 | 150 | ||
BC141056.1 | - | 12 - 13 | 1929 | 9055668 | 150 | |
CB528069.1 | - | 2 - 3 | 199 | 150 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
