Assay Details
Gene Symbol: Secisbp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177608.3 | NP_808276.2 | 15 - 16 | 2491 | 65 | |
GenBank mRNA | AK145135.1 | - | 15 - 16 | 2377 | 65 | |
AK172920.1 | - | 15 - 16 | 2358 | 65 | ||
BC071239.1 | - | 8 - 9 | 1119 | 65 | ||
BC141056.1 | - | 15 - 16 | 2451 | 9055668 | 65 | |
CB289930.1 | - | 2 - 3 | 251 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
