Assay Details
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009773.3 | NP_033903.2 | 21 - 22 | 3055 | 71 | |
GenBank mRNA | AF107296.1 | - | 21 - 22 | 3010 | 71 | |
AK088502.1 | - | 7 - 8 | 1111 | 71 | ||
AK133909.1 | - | 21 - 22 | 2996 | 71 | ||
BC031577.1 | - | 21 - 22 | 3006 | 4953223 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
