Assay Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029617.2 | NP_083893.2 | 2 - 3 | 121 | ||
GenBank mRNA | AK009772.1 | - | 2 - 3 | 121 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK009772.1, NM_029617.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
