Assay Details
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015792.1 | NP_056607.1 | 2 - 3 | 253 | 107 | |
GenBank mRNA | AK083118.1 | - | 2 - 3 | 598 | 107 | |
AK147829.1 | - | 2 - 3 | 237 | 107 | ||
AK154357.1 | - | 2 - 3 | 276 | 107 | ||
AK158937.1 | - | 2 - 3 | 304 | 107 | ||
BC031393.1 | - | 2 - 3 | 253 | 4948804 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK083118.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
